BASE Footwear Komodo Safety Shoes, Lace-Up, Low, S1P
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B0668BKR35 B0668BKR36 B0668BKR37 B0668BKR38 B0668BKR39 B0668BKR40 B0668BKR41 B0668BKR42 B0668BKR43 B0668BKR44 B0668BKR45 B0668BKR46 B0668BKR47 B0668BKR48
PRTWB0668BKR35EA 145 EUR
PRTWB0668BKR35 PRTWB0668BKR36 PRTWB0668BKR37 PRTWB0668BKR38 PRTWB0668BKR39 PRTWB0668BKR40 PRTWB0668BKR41 PRTWB0668BKR42 PRTWB0668BKR43 PRTWB0668BKR44 PRTWB0668BKR45 PRTWB0668BKR46 PRTWB0668BKR47 PRTWB0668BKR48
BASE Footwear Komodo Safety Shoes, Lace-Up, Low, S1P
Schoenen en laarzen
Komodo low safety shoes feature a Microfibre + Hi-Tech fabric upper, SmellStop anti-bacterial lining, Fresh'n Flex midsole, SlimCap toe cap, and AirTech + TPU-skin sole.
  • 200 joules resistant toe-cap
  • Puncture-resistant sole
  • Shock absorption in the heel region
  • Electronic devices protection from electrostatic discharges
  • Hydrocarbons-resistant
  • Metal-free
  • Anti-static footwear
Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces. Orthopaedic made to measure footbed, complying with european norms DGUV112-191.
Certificaten: EN ISO 20345: 2011 S1P FO ESD SR; EN 61340-4-3 S1P.