Cole-Parmer® SHM-250 Digi-Mantle Heaters
Leverancier: Cole-Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
36204-01 36204-05 36204-09 36204-13 36204-15 36204-19 36204-23
ELET36204-01EA 1470 EUR
ELET36204-01 ELET36204-05 ELET36204-09 ELET36204-13 ELET36204-15 ELET36204-19 ELET36204-23
Cole-Parmer® SHM-250 Digi-Mantle Heaters
Verwarmers Verwarmingsmantels
Heating mantles with interchangeable cartridges.
  • Set speed and temperature in seconds with the intuitive display
  • Complete system requires controller and heating cartridge
  • Achieve uniform temperature up to 450 °C across round bottom flask
  • Stirring speeds from 100 to 2000 min⁻¹ with excellent stability
  • Vented casing keeps heating cartridge cool to the touch
  • Modular design allows different size heating cartridges to be used on the same Digi-mantle controller
Control temperature and stirring speed independently on the Digi-Mantle digital controller. TheMore Product Information
Order Now
Specificaties
Elektrisch 230 VAC, 50 Hz
Environmentally Preferable
Maximumtemperatuur 450 °C