DURAN® Safety Joints, Cone with Rim
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286181402 286181908 286182407 286182904 286183403 286184502
SCOT286181402EA 94.3 EUR
SCOT286181402 SCOT286181908 SCOT286182407 SCOT286182904 SCOT286183403 SCOT286184502
DURAN® Safety Joints, Cone with Rim
Adapters Adapters met NS-verbindingen
Cones with collar and additionally moulded glass ring.
  • Made from borosilicate 3.3 glass
  • Cone with collar and additionally moulded glass ring prevents unintentional loosening of the ground joint
  • Firmly seated ground joints can be loosened more easily here, the danger of glass breakage and associated risks is thus minimised
  • Use in equipment for handling hazardous substances or working with pressure differences in the high vacuum range
  • Tight closure due to standard ground joint
In order to meet high safety demands, example when dealing with hazardous substances or pressure differences, DURAN® Safety Joints offer very good protection against accidental loosening of the connection. Furthermore, DURAN® Safety Joints offer a system to separate jammed cones and sockets. This minimises the risk of breakage and related threats.