DURAN® Measuring Cylinders
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
213970708 213970802 213971404 213971704 213972409 213973602 213974401 213975406 213976308
SCOT213970708EA 155 EUR
SCOT213970708 SCOT213970802 SCOT213971404 SCOT213971704 SCOT213972409 SCOT213973602 SCOT213974401 SCOT213975406 SCOT213976308
DURAN® Measuring Cylinders
Cilinders Maatcilinders
Class A, delivered with UKAS certificate.
  • Made from borosilicate 3.3 glass
  • Individually tested and supplied with a calibration certificate complying to the requirements of ISO 17025 and is accredited the United Kingdom Accreditation Service (UKAS)
  • The volume content tolerances conform to accuracy Class A
  • Easy-to-read scale with graduation marks to estimate the content
  • The blue printed graduations are highly chemically resistant
DURAN® volumetric measuring instruments already meet the highest accuracy standard. The DURAN® range has now been expanded to include the product range of volumetric flasks and measuring cylinders with UKAS individual certificates. This new product line is tested and calibrated according to the requirements of the United Kingdom Accreditation Services (UKAS) and complies with ISO 17025, a standard for worldwide comparability of test and calibration results.
Certificaten: Complies with ISO 4788.