BRUTE® Cleaning Accessories
Leverancier: Rubbermaid Commercial Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRUTE®
129-1894EA 116 EUR
129-1894 129-2196
BRUTE® Cleaning Accessories
Bakken Afvalbakken
Turn Rubbermaid Commercial BRUTE® waste containers into a cleaning cart with storage and transport for spray bottles, wet-floor signs, lobby dust pan, brushes, liners, gloves, and other cleaning supplies.
  • Rim caddy snaps securely onto the rim of 167 litre BRUTE® waste containers
  • Caddy bag fits 121 and 167 litre BRUTE® waste containers
The BRUTE® rim caddy measures 825×673×171 mm. BRUTE® caddy bags measure 387×286×44 mm.