Graduated Pipettes, Type 1, Class AS
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
233460606 233461105 233461602 233462307 233462907 233463209 233463406 233463603
SCOT233460606EA 94.6 EUR
SCOT233460606 SCOT233461105 SCOT233461602 SCOT233462307 SCOT233462907 SCOT233463209 SCOT233463406 SCOT233463603
Graduated Pipettes, Type 1, Class AS
Pipetten Gegradueerde pipetten
Made from soda-lime glass, delivered with certificate of conformity and batch certificate.
  • Calibrated to deliver (TD, Ex)
  • Type 1: Drain-out, zero at the top (numbering from the top down)
  • Graduation marks and inscriptions in highly durable blue print
  • With cotton plug ending (cotton plugs included)
Certificaten: Comply with ISO 835.