DURAN® Measuring Cylinders, Low Form
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
213920871 213921473 213921773 213922478 213923671 213924479 213925475 213926377
SCOT213920871EA 154 EUR
SCOT213920871 SCOT213921473 SCOT213921773 SCOT213922478 SCOT213923671 SCOT213924479 SCOT213925475 SCOT213926377
DURAN® Measuring Cylinders, Low Form
Cilinders Maatcilinders
Uncalibrated cylinders without graduation.
  • Manufactured from borosilicate 3.3 glass
  • Low shape
  • With spout and hexagonal glass base
Certificaten: Suitable for DIN EN ISO 4788.