Cole-Parmer® HM-200 Large Volume Heating Mantles
Leverancier: Cole-Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
36013-16 36013-18 36013-20 36013-22
ELET36013-16EA 5430 EUR
ELET36013-16 ELET36013-18 ELET36013-20 ELET36013-22
Cole-Parmer® HM-200 Large Volume Heating Mantles
Verwarmers Verwarmingsmantels
Accommodate 60° funnels, pear-shaped flasks and round-bottom flasks.
  • Bottom opening allows use with 60° funnels of various diameters
  • Accept large capacity flasks -between 10 to 22 litres
  • Three separate electrical circuits allow controlled heating of top, middle and bottom of flasks independently
The 'V-shaped' design of the heating mantle accommodates both pear-shaped and round bottom flasks. Coiled heating element, suspended within a thermal insulating material, provides optimal heat transfer and support. The outer casing remains cool to the touchMore Product Information
Order Now
Specificaties
Elektrisch 230 V, 50 Hz
Environmentally Preferable
Maximumtemperatuur 450 °C