WHEATON® COMPLETEPACK MODPACK Vials with Stoppers and Seals, Clear Glass
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
W991000-CPMP W991001-CPMP W991002-CPMP W991003-CPMP W991004-CPMP W991005-CPMP W991006-CPMP W991007-CPMP W991008-CPMP W991009-CPMP W991010-CPMP W991011-CPMP W991012-CPMP W991013-CPMP W991014-CPMP W991015-CPMP W991016-CPMP W991017-CPMP
WHEAW991000-CPMPEA 19190 EUR
WHEAW991000-CPMP WHEAW991001-CPMP WHEAW991002-CPMP WHEAW991003-CPMP WHEAW991004-CPMP WHEAW991005-CPMP WHEAW991006-CPMP WHEAW991007-CPMP WHEAW991008-CPMP WHEAW991009-CPMP WHEAW991010-CPMP WHEAW991011-CPMP WHEAW991012-CPMP WHEAW991013-CPMP WHEAW991014-CPMP WHEAW991015-CPMP WHEAW991016-CPMP WHEAW991017-CPMP
WHEATON® COMPLETEPACK MODPACK Vials with Stoppers and Seals, Clear Glass
flesjes
Six inventory matched kits of clear crimp top vials, rubber stoppers and seals that are sterilised and ready-to-use packed into a shipper case.
  • Sterilised, off-the-shelf, ready-to-use (RtU) primary packaging components
  • Complete manufacturing and certificate of analysis traceability
  • Ready-to-use, just-in-time, minimum 180 day shelf life from time of shipment
  • Container Closure Integrity (CCI) validation
Certificaten: USP <788>/EP 2.9.19 Particulate Matter in Injections (Vials and Stoppers), USP <85>/EP 2.6.1, The Bacterial Endotoxins Test (Vials and Stoppers) and lot certified to SAL 10⁻⁶ (Vials, Stoppers, and Seals), USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations.