Chemical Spill Kits
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HSRK5 SK210 SK75N DRSK_DP
MMMAHSRK5EA 0 EUR
MMMAHSRK5 MMMASK210 MMMASK75N MMMADRSK_DP
Chemical Spill Kits
Absorptiematerialen en bescherming tegen morsen
The 3M™ Hazardous spill response Kits help make sure you are ready with everything you need should a spill occur.
  • Ideal for rapid response to chemical spills
  • Kits allow fast and effective containment and clean-up
  • Clean up hazardous chemical spills, drips, and leaks
  • Suitable for use with most chemicals
  • Easy disposal
Kits assist in cleaning up hazardous chemical spills quickly in an emergency situation. A selection of chemical sorbents of varying formats are included, as are clean-up bags.