Scotch-Brite™ 3001HEX Low-Scratch Scouring Sponge
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Scotch-Brite™
MMMA3001HEXEA 28.5 EUR
MMMA3001HEX
Scotch-Brite™ 3001HEX Low-Scratch Scouring Sponge
Schoonmaakdoekjes Sponzen
The Scotch-Brite™ Low scratch scouring sponge 3001HEX is an effective two-in-one scouring sponge. It features a blue power dot side that’s engineered to quickly and effectively power through tough messes with less visible scratches. The reverse foam sponge side is designed to wipe surfaces clean.
  • Clean a wide variety of surfaces where minimal scratching is desired
  • Ergonomic hexagon shape fits comfortably in the hand
  • Easy to clean hard to reach areas
  • HACCP International certified
Clean a wide variety of surfaces with minimal scratches with Scotch-Brite™ Low scratch scouring sponge 3001HEX. The sponge features a special hexagon design that makes it easy to get in hard-to-reach areas of pots and pans and the shape fits comfortably in hand, maximizing productivity and minimizing operator fatigue. The blue power dot side is engineered to quickly and effectively power through tough messes with less visible scratches, while the foam sponge side wipes surface clean.