Water Dissolvable Paper Labels for M610, M611, and BMP61
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173534 173536
BRDY173534EA 114.6 EUR
BRDY173534 BRDY173536
Water Dissolvable Paper Labels for M610, M611, and BMP61
Etiketten
This B-403 material completely dissolves in warm water within 30 seconds, leaving no adhesive residue behind.
  • Ideal for use in laboratory settings, asset tracking, or general use
  • Easy to remove, environmentally responsible, print-on-demand THT label solution
  • Easy removal saves time during re-labelling processes
  • Pressure-sensitive adhesive allows for quick and easy application
  • Matte finish
Applications include asset tracking, general identification, and laboratory labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310.
Verpakking: One tape per box.