Repositionable Multi-Purpose Vinyl Labels for M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173288 173221 173258 173257 173289 173222
BRDY173288EA 78.89 EUR
BRDY173288 BRDY173221 BRDY173258 BRDY173257 BRDY173289 BRDY173222
Repositionable Multi-Purpose Vinyl Labels for M710 and BMP71
Etiketten
Repositionable vinyl film (B-581) is a durable, flexible, high gloss material ideal for indoor marking where frequent repositioning is required.
  • Special aggressive adhesive stays in place but removes cleanly without leaving adhesive residue
  • Ideal for applications like 5S and inventory
  • Vinyl film can be thermal transfer printed
  • Gloss finish
  • Average outdoor durability is five years
  • Service temperature of −40 to +82 °C (−40 to +180 °F)
Labels are ideal for facility identification, general identification, and safety identification applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
Verpakking: One roll per box.