Ultra Aggressive Adhesive Multi-Purpose Polyester Labels for M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173416 173314 173407 173328 173294 173439 173359 174623 173388 173441 174639 173389 174577 174667 173387 173436 173437 173342 173435 173392 174624 173397 173380 173391 173390 173350 173440 174622 173324 173438 174625 173265 173393
BRDY173416EA 66.52 EUR
BRDY173416 BRDY173314 BRDY173407 BRDY173328 BRDY173294 BRDY173439 BRDY173359 BRDY174623 BRDY173388 BRDY173441 BRDY174639 BRDY173389 BRDY174577 BRDY174667 BRDY173387 BRDY173436 BRDY173437 BRDY173342 BRDY173435 BRDY173392 BRDY174624 BRDY173397 BRDY173380 BRDY173391 BRDY173390 BRDY173350 BRDY173440 BRDY174622 BRDY173324 BRDY173438 BRDY174625 BRDY173265 BRDY173393
Ultra Aggressive Adhesive Multi-Purpose Polyester Labels for M710 and BMP71
Etiketten
ToughBond® ultra aggressive polyester (B-483) is perfect for labelling products and general ID and can be applied to powder coated, smooth, textured, or irregular surfaces.
  • Glossy white polyester with permanent rubber based adhesive is thermal transfer printable
  • Material is designed for high adhesion to textured metals and low surface energy plastics
  • Service temperature of −40 to +120 °C (−40 to +248 °F)
Labels are ideal for barcode labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, generalMore Product Information