Aggressive Adhesive Multi-Purpose Polyester Labels for M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173320 173234 173203 173229 173298 173330 173231 173267
BRDY173320EA 65.1 EUR
BRDY173320 BRDY173234 BRDY173203 BRDY173229 BRDY173298 BRDY173330 BRDY173231 BRDY173267
Aggressive Adhesive Multi-Purpose Polyester Labels for M710 and BMP71
Etiketten
Thermal transfer printable, glossy permanent polyester (B-422) labels are ideal for use in all commercial and industrial applications.
  • Aggressive adhesive good for powder-coated and textured flat surfaces, including low surface energy plastics
  • Designed to withstand exposure to numerous solvents
  • Gloss finish
  • Service temperature of −40 to +100 °C (−40 to +212 °F)
Labels are ideal for barcode labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, inventory and inspection labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
Certificaten: UL recognised. CSA accepted.