Aggressive Adhesive Multi-Purpose Nylon Labels B-499 for M610, M611, BMP61 Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174155 174103 174151 174077 174085 174116 174165 174086
BRDY174155EA 33.12 EUR
BRDY174155 BRDY174103 BRDY174151 BRDY174077 BRDY174085 BRDY174116 BRDY174165 BRDY174086
Aggressive Adhesive Multi-Purpose Nylon Labels B-499 for M610, M611, BMP61 Printers
Etiketten
Nylon cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as vials.
  • Service temperature of −40 to +90 °C (−40 to +193 °F)
  • Low-profile material adheres to surfaces that are smooth, rough, or powder-coated
  • Also adheres to irregular surfaces or surfaces with low surface energy
  • Matte finish
Ideal for component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, laboratory labelling, and wire and cable labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310.
Certificaten: UL recognised. CSA accepted. Halogen-free (DIN VDE 0472/part 815). Approvals are for white material only.