ToughWash® Metal Detectable Washdown Resistant Labels for M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173469 173459 173468 173458
BRDY173469EA 294.29 EUR
BRDY173469 BRDY173459 BRDY173468 BRDY173458
ToughWash® Metal Detectable Washdown Resistant Labels for M710 and BMP71
Etiketten
ToughWash® metal detectable polyester film with acrylic adhesive (B-854) is a strong, print on demand label specially formulated for the food and beverage processing industry.
  • Capable of being detected by commercially available metal detectors
  • Label construction is tough against harsh washdown environments
  • Will withstand washdowns up to 80 °C (175 °F) and pressure up to 103,4 bar
  • Service temperature of −40 to +110 °C (−40 to +230 °F)
  • Matte finish
Ideal for facility identification, food and beverage process labelling, general identification, and safety identification applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
Verpakking: One roll per box.