Metallised Matte Gray Polyester Labels for M610, M611, BMP61, M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174433 174457
BRDY174433EA 525.12 EUR
BRDY174433 BRDY174457
Metallised Matte Gray Polyester Labels for M610, M611, BMP61, M710 and BMP71
Etiketten
Metallised Polyester (B-428) labels are designed for applications that use barcodes, alphanumerics, graphic symbols and logos, and require nameplate-like quality.
  • Matte, light gray finish gives labels a rating and serial plate appearance
  • Labels are designed to withstand numerous solvents and variable temperatures when applied to various surfaces
  • Service temperature of −40 to +120 °C (−40 to +248 °F)
Ideal for asset tracking, circuit board labelling, component and equipment labelling, data and telecommunications labelling, general identification, inventory and inspection labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R4300 for the M710/BMP71 label printers.
Certificaten: UL recognised. CSA accepted. AGA approved. Halogen-free (DIN VDE 0472/part 815).