Aggressive Adhesive Multi-Purpose Polyester Labels for M610, M611 and BMP61
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174152 174164
BRDY174152EA 54.35 EUR
BRDY174152 BRDY174164
Aggressive Adhesive Multi-Purpose Polyester Labels for M610, M611 and BMP61
Etiketten
Thermal transfer printable, glossy permanent polyester (B-422) labels are ideal for use in all commercial and industrial applications.
  • Aggressive adhesive good for powder-coated and textured flat surfaces, including low surface energy plastics
  • Designed to withstand exposure to numerous solvents
  • Gloss finish
  • Service temperature of −40 to 100° C (−40 to 212 °F)
Ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, and inventory and inspection labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010.
Certificaten: UL recognised. CSA accepted.