Aggressive Adhesive Multi-Purpose Polyester Labels B-422 for M610, M611, BMP61, M710 (with Media Adapter), BMP71 (with Media Adapter)
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173961 174331 174327 174206 174266 174298 174196 174304 174227 174291 173967 174333 174239 174359 174255 173964 174329 174306 174344 173969 174219 173758 174316 173831
BRDY173961EA 85.92 EUR
BRDY173961 BRDY174331 BRDY174327 BRDY174206 BRDY174266 BRDY174298 BRDY174196 BRDY174304 BRDY174227 BRDY174291 BRDY173967 BRDY174333 BRDY174239 BRDY174359 BRDY174255 BRDY173964 BRDY174329 BRDY174306 BRDY174344 BRDY173969 BRDY174219 BRDY173758 BRDY174316 BRDY173831
Aggressive Adhesive Multi-Purpose Polyester Labels B-422 for M610, M611, BMP61, M710 (with Media Adapter), BMP71 (with Media Adapter)
Etiketten
Thermal transfer printable, glossy permanent polyester (B-422) labels are ideal for use in all commercial and industrial applications.
  • Aggressive adhesive good for powder-coated and textured flat surfaces, including low surface energy plastics
  • Designed to withstand exposure to numerous solvents
  • Service temperature of −40 to 100 °C (−40 to 212 °F)
  • Gloss finish
Ideal for barcode labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, inventory and inspection labellingMore Product Information