Electrostatic Dissipative (ESD) Polyimide Labels for M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174552 174592
BRDY174552EA 218.46 EUR
BRDY174552 BRDY174592
Electrostatic Dissipative (ESD) Polyimide Labels for M710 and BMP71
Etiketten
Static Dissipative Polyimide (B-717) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labelling.
  • High temperature, glossy white polyimide material
  • Static dissipative adhesive and static dissipative release liner
  • Surface resistivity values in the recommended range for dissipative ESD packaging materials as defined by ANSI/ESD S541-2008
  • Service temperature of −70 to 100 °C (−94 to 212 °F)
Ideal for circuit board labelling.
Brady Authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
Certificaten: UL recognised. Meets the requirements of ANSI/ESD S541-2008 (between 10⁴ and 10¹ ohms) for dissipative ESD packaging materials. Meets the performance requirements of MIL-STD-202G, Method 215K.