Harsh Environment Multi-Purpose Polyester Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
152223 174228 174256 174246 174242 174299 174334 174249 174190 174244 174245 174247 174345 174186 174292 174240 174250 174337 174197 174287 174326 174189 174372 174360 174282 174173 174193 174207 174274
BRDY152223EA 65.79 EUR
BRDY152223 BRDY174228 BRDY174256 BRDY174246 BRDY174242 BRDY174299 BRDY174334 BRDY174249 BRDY174190 BRDY174244 BRDY174245 BRDY174247 BRDY174345 BRDY174186 BRDY174292 BRDY174240 BRDY174250 BRDY174337 BRDY174197 BRDY174287 BRDY174326 BRDY174189 BRDY174372 BRDY174360 BRDY174282 BRDY174173 BRDY174193 BRDY174207 BRDY174274
Harsh Environment Multi-Purpose Polyester Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Etiketten
Permanent polyester (B-423) is ideal for electronic PCB and component identification, bar code labels, rating plates, and solar panel identification.
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • Service temperature of −70 to 110 °C (−94 to 230 °F)
Ideal for barcode labelling, circuit board labelling, component and equipment labelling, general identification, and inventory and inspection labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R6000 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Certificaten: UL recognised. CSA accepted. Halogen-free (DIN VDE 0472/part 815). Approvals are for White material only.