FreezerBondz™ Cryogenic Matte Polyester Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173708 174179 173709 174352 173904
BRDY173708EA 51.77 EUR
BRDY173708 BRDY174179 BRDY173709 BRDY174352 BRDY173904
FreezerBondz™ Cryogenic Matte Polyester Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Etiketten
Durable Polyester (B-490) markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws, and slides.
  • White polyester material has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance and solvent resistance
  • High/low temperature performance of −196 to 100 °C (−320 to 212 °F)
  • Matte finish
Ideal for laboratory labelling applications. B-490 labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen when wrappedMore Product Information