Repositionable Multi-Purpose Vinyl Cloth Labels B-498 CleanLift® for M610, M611, BMP61, M710 (with media adapter), BMP71 (with media adapter) Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173851 174224 173860 174214 174237 174279 174195 174183 174264 174348 174272 174176 174217 173768 173789
BRDY173851EA 72.15 EUR
BRDY173851 BRDY174224 BRDY173860 BRDY174214 BRDY174237 BRDY174279 BRDY174195 BRDY174183 BRDY174264 BRDY174348 BRDY174272 BRDY174176 BRDY174217 BRDY173768 BRDY173789
Repositionable Multi-Purpose Vinyl Cloth Labels B-498 CleanLift® for M610, M611, BMP61, M710 (with media adapter), BMP71 (with media adapter) Printers
Etiketten
Permanent or repositionable, once applied the B-498 CleanLift® Series Vinyl Cloth will remain in place until change is required.
  • Adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning
  • Sticks to any clean, dry surface
  • Specially formulated topcoat for excellent print quality
  • Semi-gloss finish for crisp, clean look
Ideal for barcode labelling, circuit board labelling, component and equipment labelling, laboratory labelling, and wire and cable labelling applications.
Brady authentic materials workMore Product Information