Electrostatic Dissipative (ESD) Polyimide Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174290 174203 174374 173775 174177 174191 173778 173773
BRDY174290EA 155.85 EUR
BRDY174290 BRDY174203 BRDY174374 BRDY173775 BRDY174177 BRDY174191 BRDY173778 BRDY173773
Electrostatic Dissipative (ESD) Polyimide Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Etiketten
Static Dissipative Polyimide (B-717) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labelling.
  • High temperature, glossy white polyimide material
  • Static dissipative adhesive and static dissipative release liner
  • Surface resistivity values in the recommended range for Dissipative ESD packaging materials
  • Service temperature of −70 to 100 °C (−94 to 212 °F)
Ideal for circuit board labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R6000 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Certificaten: UL recognised. Meets the requirements of ANSI/ESD S541-2008 (between 10⁴ and 10¹¹ ohms) for dissipative ESD packaging materials. Meets the performance requirements of MIL-STD-202G, Method 215K.