Harsh Environment Multi-Purpose Clear Polyester Labels for M610, M611 and BMP61
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174100 174102 174105 174106
BRDY174100EA 63.68 EUR
BRDY174100 BRDY174102 BRDY174105 BRDY174106
Harsh Environment Multi-Purpose Clear Polyester Labels for M610, M611 and BMP61
Etiketten
Clear Polyester (B-430) labels are designed for applications that use barcodes, alphanumerics, graphic symbols and logos, and require nameplate-like quality.
  • Glossy, clear labels are thermal transfer printable and feature a durable permanent acrylic adhesive
  • Excellent resistance to most chemicals, oils, and water
  • Service temperature of −56 to 137 °C (−70 to 100 °F)
Ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, and inventory and inspection labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010.
Certificaten: UL recognised. CSA accepted.