Ultra-Thin Matte Electrostatic Dissipative (ESD) Polyimide Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173883 173712 173919 173722 173896 173731 173698 173793
BRDY173883EA 171.67 EUR
BRDY173883 BRDY173712 BRDY173919 BRDY173722 BRDY173896 BRDY173731 BRDY173698 BRDY173793
Ultra-Thin Matte Electrostatic Dissipative (ESD) Polyimide Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Etiketten
Static Dissipative Polyimide (B-719) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labelling.
  • Low profile, high temperature, matte white polyimide material
  • Static dissipative adhesive and static dissipative release liner
  • Surface resistivity values in the recommended range for Dissipative ESD packaging materials
  • Service temperature of −70 to 100 °C (−94 to 212 °F)
Ideal for circuit board labelling.
Brady authentic materials work seamlessly with Brady printers for improvedMore Product Information