Ultra Aggressive Adhesive Metallised Matte Polyester Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174234 174262 174302 174213 174271 174223 174278 174296 173718 173842 173772
BRDY174234EA 51.55 EUR
BRDY174234 BRDY174262 BRDY174302 BRDY174213 BRDY174271 BRDY174223 BRDY174278 BRDY174296 BRDY173718 BRDY173842 BRDY173772
Ultra Aggressive Adhesive Metallised Matte Polyester Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Etiketten
Ultra Aggressive Metallised Polyester (B-486) is ideal for rating/serial plates that use barcodes, alphanumerics, graphic symbols and logos, and require nameplate-like quality.
  • Matte, light gray material has a permanent rubber-based adhesive and is thermal transfer printable
  • Designed for high adhesion to textured metals, powder-coated surfaces and low energy plastics
  • Service temperature of −40 to 120 °C (−40 to 248 °F)
Ideal for asset tracking, component and equipment labelling, general identification, and inventory and inspection labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R4300 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Certificaten: UL recognised. CSA accepted.