Electrostatic Dissipative (ESD) Polyester Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174252 174211 174192 174175 174339 174373 174199 173886
BRDY174252EA 158.5 EUR
BRDY174252 BRDY174211 BRDY174192 BRDY174175 BRDY174339 BRDY174373 BRDY174199 BRDY173886
Electrostatic Dissipative (ESD) Polyester Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Etiketten
This B-473 material is a low profile polyimide and polyester material that has static dissipative properties.
  • Primarily used for printed circuit boards and electronic component labelling
  • B-473 material is a white glossy polyester with a permanent static dissipative adhesive and liner
  • Surface resistivity values in the recommended range for dissipative ESD packaging materials
  • Abrasive-resistant
  • Service temperature of −40 to 120 °C (−40 to 248 °F)
Ideal for use in printed circuit board and electronic component post-process labelling.
Brady authenticMore Product Information