Harsh Environment Multi-Purpose Polyester Labels for M610, M611 and BMP61
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174099 174104 173805 173806 173804
BRDY174099EA 55.78 EUR
BRDY174099 BRDY174104 BRDY173805 BRDY173806 BRDY173804
Harsh Environment Multi-Purpose Polyester Labels for M610, M611 and BMP61
Etiketten
Permanent polyester (B-423) is ideal for electronic PCB and component identification, bar code labels, rating plates, and solar panel identification.
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • Service temperature of −70 to 110 °C (−94 to 230 °F)
Ideal for barcode labelling, circuit board labelling, component and equipment labelling, general identification, and inventory and inspection labelling applications. Brady B­423 is also UL recognised for outdoor use on glass, thermoset polyester plastic and polyvinyl fluorideMore Product Information