Self-Laminating Self-Extinguishing Tedlar Wrap Around Label for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174226 174238 174281
BRDY174226EA 91.22 EUR
BRDY174226 BRDY174238 BRDY174281
Self-Laminating Self-Extinguishing Tedlar Wrap Around Label for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Etiketten
The B-642 is a thin tape with self-extinguishing properties that has excellent flexibility for wrapping on small diameter wires.
  • Ideal for use in aerospace, defense and mass transit industries
  • Polyvinylfluoride film provides good self-extinguishing properties and solvent resistance
  • For use in dot matrix and thermal transfer printers
  • Matte finish
Ideal for data and telecommunications labelling, electrical labelling, and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R4300 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Certificaten: RoHS Compatibility Pass 2011/65/EU.