Polyester Die-cut Labels for M610/M611 Printer
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173501 173531 173499 173490 173530 173528 173493 173500 173492 173489 173529 173503 173532 173504 173502 173527 173494 173491
BRDY173501EA 88.37 EUR
BRDY173501 BRDY173531 BRDY173499 BRDY173490 BRDY173530 BRDY173528 BRDY173493 BRDY173500 BRDY173492 BRDY173489 BRDY173529 BRDY173503 BRDY173532 BRDY173504 BRDY173502 BRDY173527 BRDY173494 BRDY173491
Polyester Die-cut Labels for M610/M611 Printer
Etiketten
The B-494 polyester label is a reliable sample identification and traceability solution. The label will stay attached and remain legible, even when exposed to chemicals or stored in liquid nitrogen.
  • Color printed below the label surface withstands harsh lab environments
  • Will withstand liquid nitrogen, hot water baths and more
  • Resistant to common lab chemicals like ethanol and xylene when used with the R6400 ribbon series
  • Gloss finish
Thanks to subsurface printing, the label's colour code will not fade, smear or smudge and will continue to offer fastMore Product Information