Self-laminating Vinyl Labels for M611, M610, and M710
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174257 174285 174180 174236 174288 174208 174216 174283 174243 174275 173894
BRDY174257EA 71.36 EUR
BRDY174257 BRDY174285 BRDY174180 BRDY174236 BRDY174288 BRDY174208 BRDY174216 BRDY174283 BRDY174243 BRDY174275 BRDY173894
Self-laminating Vinyl Labels for M611, M610, and M710
Etiketten
Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text.
  • Durable, permanent acrylic adhesive has outstanding water and oil resistance
  • Service temperature of −40 to +70 °C (−40 to +158 °F)
  • Matte finish
Labels are ideal for data and telecommunications labelling, electrical labelling, laboratory labelling, and wire and cable labelling applications.
Labels for the M610, M611, and M710 portable printers. A ribbon is required to print this material. Use part number M61-R4310 for the M610 label printer/Brady printer M611 mobile label printer or M7-R4300 for the M710 label printer.
Certificaten: UL recognised to UL969 labelling and marking standard when printed with the Brady series R4300 and R6200 ribbons. See UL files MH17154 for specific details. Brady customers should specify at purchase if they require UL recognised B-427 labels. Brady sells both UL recognised B-427 labels and non-approved labels depending on customer requirements.