Self-Laminating Vinyl Wrap Around Labels for M610, M611, BMP61, M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173442 173447 173449 174412 173453 173450 173454 173452 174492 173451 174432 173446 173455 174469 174411
BRDY173442EA 354.61 EUR
BRDY173442 BRDY173447 BRDY173449 BRDY174412 BRDY173453 BRDY173450 BRDY173454 BRDY173452 BRDY174492 BRDY173451 BRDY174432 BRDY173446 BRDY173455 BRDY174469 BRDY174411
Self-Laminating Vinyl Wrap Around Labels for M610, M611, BMP61, M710 and BMP71
Etiketten
Self-laminating vinyl (B-427) is an excellent material for wire and cable identification. This material has good clarity and conformability, and is self-extinguishing.
  • Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text
  • Durable, permanent acrylic adhesive has outstanding water and oil resistance
  • Service temperature of −40 to +70 °C (−40 to +158 °F)
  • Matte finish
Ideal for data and telecommunications labelling, electrical labelling, laboratory labelling, and wire and cable labellingMore Product Information
Order Now
Specificaties
Environmentally Preferable