Self-Laminating Cryogenic Polyester Labels for M610, M611 and BMP61
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174079 174080 174091 174149 174092 174153 174093
BRDY174079EA 77.35 EUR
BRDY174079 BRDY174080 BRDY174091 BRDY174149 BRDY174092 BRDY174153 BRDY174093
Self-Laminating Cryogenic Polyester Labels for M610, M611 and BMP61
Etiketten
Self-laminating polyester (B-461) is a clear film with a matte white printable zone ideal for cables, laboratory ID, or general labelling where graphics need to be protected.
  • Good smudge resistance, solvent resistance, and high/low-temperature performance
  • Service temperature of −196 to +110 °C (−320 to +230 °F)
  • Self-laminated label with a white printable zone and translucent overlaminating area to protect text
  • Matte finish
Ideal for component and equipment labelling, general identification, and laboratory labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310.