Indoor/Outdoor Vinyl Labels for M710
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173271 173272
BRDY173271EA 82.11 EUR
BRDY173271 BRDY173272
Indoor/Outdoor Vinyl Labels for M710
Etiketten
B-595 vinyl will adhere to difficult low surface energy items, including PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics. The B-595 material also excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes.
  • Permanent acrylic adhesive keeps labels adhered in harsh climates and is resistant to chemicals, oils and detergents
  • Vinyl tape is ideal in both indoor and outdoor use
  • Adheres to surfaces that are smooth, rough or powder coated, and to irregular surfaces or surfaces with low surface energy
  • Prints on white vinyl background with a clean gloss finish that is resistant to exposure to chemicals, grease, oil, and cleaning agents
  • Average outdoor durability of 8-10 years in temperatures ranging from −40 to 82 °C (−40 to +180 °F)
  • Gloss finish
Ideal for component and equipment labelling, electrical labelling, facility identification, floor marking, laboratory labelling, lean and 5S labelling, pipe and valve marking, and warehouse marking applications.
A ribbon is required to print this material. Use part number M7-R10000.
Certificaten: BS5609:1986 section 2 and section 3.
Verpakking: 1 roll per box.