FreezerBondz™ Ultra-Thin Cryogenic Polyester Labels for M610, M611 and BMP61
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174082 173481 174089 174097 174107 174096 174087 174084 174154 174110 174095 174076 174114
BRDY174082EA 100.62 EUR
BRDY174082 BRDY173481 BRDY174089 BRDY174097 BRDY174107 BRDY174096 BRDY174087 BRDY174084 BRDY174154 BRDY174110 BRDY174095 BRDY174076 BRDY174114
FreezerBondz™ Ultra-Thin Cryogenic Polyester Labels for M610, M611 and BMP61
Etiketten
Durable FreezerBondz™ polyester (B-492) markers are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
  • Can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Good print smudge resistance and solvent resistance
  • High/low-temperature performance range of −196 to +100 °C (−320 to +212 °F)
  • Matte finish
Ideal for component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, and laboratoryMore Product Information