Metallised Matte Grey Polyester Labels for M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173325 174576 173410 173352 173420 173382 173227 173368 173361
BRDY173325EA 75.52 EUR
BRDY173325 BRDY174576 BRDY173410 BRDY173352 BRDY173420 BRDY173382 BRDY173227 BRDY173368 BRDY173361
Metallised Matte Grey Polyester Labels for M710 and BMP71
Etiketten
Metallised polyester (B-428) labels are designed for applications that use barcodes, alphanumerics, graphic symbols, and logos and require nameplate-like quality.
  • Matte, light grey finish gives labels a rating and serial plate appearance
  • Labels are designed to withstand numerous solvents and variable temperatures when applied to various surfaces
  • Service temperature of −40 to +120 °C (−40 to +248 °F)
Ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, and inventory and inspection labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R4300.
Certificaten: UL recognised. CSA accepted. AGA approved. Halogen-free (DIN VDE 0472/part 815).