Harsh Environment Multi-Purpose Polyester Labels for M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173384 173371 173317 173228 173329 173363 173412 173304 173370 173419 174595 173318 173422 173313 174590 173434 173374 173423 173366 173338 173369 173400 173425 173373 173367 173421 173326 173333 173321 173346
BRDY173384EA 32.7 EUR
BRDY173384 BRDY173371 BRDY173317 BRDY173228 BRDY173329 BRDY173363 BRDY173412 BRDY173304 BRDY173370 BRDY173419 BRDY174595 BRDY173318 BRDY173422 BRDY173313 BRDY174590 BRDY173434 BRDY173374 BRDY173423 BRDY173366 BRDY173338 BRDY173369 BRDY173400 BRDY173425 BRDY173373 BRDY173367 BRDY173421 BRDY173326 BRDY173333 BRDY173321 BRDY173346
Harsh Environment Multi-Purpose Polyester Labels for M710 and BMP71
Etiketten
Permanent polyester (B-423) is ideal for electronic PCB and component identification, bar code labels, rating plates, and solar panel identification.
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • Meets requirements of a halogen-free material per DIN VDE 0472 part 815
  • Service temperature of −70 to +110 °C (−94 to +230 °F)
Ideal for barcode labelling, circuit board labelling, component and equipment labelling, general identification, inventory and inspection labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
Certificaten: UL recognised. CSA accepted. Halogen-free (DIN VDE 0472/part 815). Approvals are for white material only.