Repositionable Multi-Purpose Vinyl Cloth Labels B-498 CleanLift® for M610, M611, BMP61, M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173476 174434 174454 174416 174470
BRDY173476EA 391.3 EUR
BRDY173476 BRDY174434 BRDY174454 BRDY174416 BRDY174470
Repositionable Multi-Purpose Vinyl Cloth Labels B-498 CleanLift® for M610, M611, BMP61, M710 and BMP71
Etiketten
Permanent or repositionable, once applied, B-498 CleanLift® series vinyl cloth remains in place, until a change is required.
  • Adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning
  • Sticks to any clean, dry surface
  • Specially formulated topcoat for excellent print quality
  • Semi-gloss finish for crisp, clean look
Ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6210 for the M610/BMP61/BradyPrinter M611/M611 Label Printers or M7-R6200 for the M710/BMP71 Label Printers.
Certificaten: UL recognised.