Repositionable Multi-Purpose Vinyl Cloth Labels B-498 CleanLift® for M610, M611, BMP61 Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174143 174146 174150 174075 173913
BRDY174143EA 61.02 EUR
BRDY174143 BRDY174146 BRDY174150 BRDY174075 BRDY173913
Repositionable Multi-Purpose Vinyl Cloth Labels B-498 CleanLift® for M610, M611, BMP61 Printers
Etiketten
Permanent or repositionable, once applied, B-498 CleanLift® series vinyl cloth remains in place, until a change is required.
  • Adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning
  • Sticks to any clean, dry surface
  • Specially formulated topcoat for excellent print quality
  • Semi-gloss finish for crisp, clean look
Ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6210.
Certificaten: UL recognised.