T-Flag or P-Flag Matte Polypropylene Labels for M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173201 174733 173200
BRDY173201EA 58.54 EUR
BRDY173201 BRDY174733 BRDY173200
T-Flag or P-Flag Matte Polypropylene Labels for M710 and BMP71
Etiketten
White B-425 thermal transfer printable polypropylene is ideal for labelling applications requiring excellent solvent resistance and print performance.
  • No visible affect to label when exposed to most common chemicals, oils, or water
  • Service temperature of −70 to +80 °C (−94 to +176 °F)
  • Matte finish
Ideal for asset tracking, barcode labelling, component and equipment labelling, data and telecommunications labelling, general identification, inventory and inspection labelling, laboratory labelling, and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R4300.
Certificaten: UL recognised. CSA accepted. Halogen-free (DIN VDE 0472/part 815).