Matte Amber Ultra Heat Resistant Polyimide Labels for M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174555 173302 173404 173424 173364
BRDY174555EA 221.08 EUR
BRDY174555 BRDY173302 BRDY173404 BRDY173424 BRDY173364
Matte Amber Ultra Heat Resistant Polyimide Labels for M710 and BMP71
Etiketten
Amber high temperature polyimide with an ultra-durable adhesive withstands the extreme wash protocol and cleaning chemicals used in printed circuit board manufacturing.
  • Withstands multiple harsh condition wash cycles for printed circuit boards and electronic component preprocess labelling
  • Resists high temperatures up to 300 °C (572 °F)
  • Survives on the top or bottom side of printed circuit boards through the solder process
  • Matte finish
Ideal for circuit board labelling and component and equipment labelling.
Brady authentic materials work seamlesslyMore Product Information