Harsh Environment Multi-Purpose Clear Polyester Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174362 173875 174346 174198 174332 174210 174328 174307 174330 174317 174251 173893 173898
BRDY174362EA 61.61 EUR
BRDY174362 BRDY173875 BRDY174346 BRDY174198 BRDY174332 BRDY174210 BRDY174328 BRDY174307 BRDY174330 BRDY174317 BRDY174251 BRDY173893 BRDY173898
Harsh Environment Multi-Purpose Clear Polyester Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Etiketten
Clear polyester (B-430) labels are designed for applications that use barcodes, alphanumerics, graphic symbols, and logos and require nameplate-like quality.
  • Glossy, clear labels are thermal transfer printable and feature a durable permanent acrylic adhesive
  • Excellent resistance to most chemicals, oils and water
  • Service temperature of −56 to +137 °C (−70 to +100 °F)
Ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, general identification, inventory and inspection labelling, and laboratory labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010 for the M610/BMP61/BradyPrinter M611/M611 Label Printers or M7-R6000 for the M710/BMP71 Label Printers. This label is no longer compatible with TLS series printers.
Certificaten: UL recognised. CSA accepted.