Electrostatic Dissipative (ESD) Polyester Labels for M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173408 173365 173417 173432 173303 173315
BRDY173408EA 101.01 EUR
BRDY173408 BRDY173365 BRDY173417 BRDY173432 BRDY173303 BRDY173315
Electrostatic Dissipative (ESD) Polyester Labels for M710 and BMP71
Etiketten
This B-473 material is a low profile polyimide and polyester material that has static dissipative properties.
  • Primarily used for printed circuit boards and electronic component labelling
  • The B-473 material is a white glossy polyester with a permanent static dissipative adhesive and liner
  • Has surface resistivity values in the recommended range for dissipative ESD packaging materials
  • Gloss finish
Ideal for circuit board labelling, electrical labelling, and general identification applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
Certificaten: UL recognised. CSA accepted. Meets surface resistivity values in the recommended range for dissipative ESD packaging materials as defined by ANSI/ESD S541-2008 (between 10⁴ and 10¹¹ ohms).