Harsh Environment Multi-Purpose Polyester Labels for M610, M611, BMP61, M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174453 173448 174493 174468 174431 173445
BRDY174453EA 285.99 EUR
BRDY174453 BRDY173448 BRDY174493 BRDY174468 BRDY174431 BRDY173445
Harsh Environment Multi-Purpose Polyester Labels for M610, M611, BMP61, M710 and BMP71
Etiketten
Permanent polyester (B-423) is ideal for electronic PCB and component identification, bar code labels, rating plates, and solar panel identification.
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • Service temperature of −70 to +110 °C (−94 to +230 °F)
Ideal for barcode labelling, circuit board labelling, component and equipment labelling, general identification, inventory and inspection labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010 for the M610/BMP61/BradyPrinter M611/M611 Label Printers or M7-R6000 for the M710/BMP71 Label Printers.
Certificaten: UL recognised. CSA accepted. Halogen-free (DIN VDE 0472/part 815). Approvals are for white material only.