Ultra Aggressive Adhesive Multi-Purpose Matte Polyester Labels for M710 and BMP71
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
173358 173323 174558 173336
BRDY173358EA 89.65 EUR
BRDY173358 BRDY173323 BRDY174558 BRDY173336
Ultra Aggressive Adhesive Multi-Purpose Matte Polyester Labels for M710 and BMP71
Etiketten
Ultra aggressive polyester (B-489A) with permanent adhesive is ideal for labelling products and general ID in both commercial and industrial applications.
  • Best for application on smooth and highly textured indoor surfaces, including metal and low surface energy plastics
  • Matte white polyester material has a permanent rubber based adhesive and is thermal transfer printable
  • Service temperature of −40 to +120 °C (−40 to +248 °F)
Ideal for component and equipment labelling, data and telecommunications labelling, and general identification applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R4300.
Certificaten: UL recognised. CSA accepted.