StainerBondz™ Microscope Slide Matte Polyester Labels for M610, M611 and BMP61
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174090 174113 174088
BRDY174090EA 65.65 EUR
BRDY174090 BRDY174113 BRDY174088
StainerBondz™ Microscope Slide Matte Polyester Labels for M610, M611 and BMP61
Etiketten
Super chemical-resistant polyester (B-481) is designed for use in the slide staining process and other laboratory ID which requires superior chemical resistance.
  • Performs well in the commonly used H&E (hematoxilin and eosin) slide staining process
  • Excellent resistance to a wide variety of chemicals, oils, and water
  • Service temperature of −80 to +130 °C (−112 to +266 °F)
Ideal for healthcare and hospital labelling, general identification, and laboratory labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6410.