Metallised Matte Grey Polyester Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174268 174258 174294 174230 174248 174194 174361 174300 174209 173757 173770
BRDY174268EA 90.36 EUR
BRDY174268 BRDY174258 BRDY174294 BRDY174230 BRDY174248 BRDY174194 BRDY174361 BRDY174300 BRDY174209 BRDY173757 BRDY173770
Metallised Matte Grey Polyester Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Etiketten
Metallised polyester (B-428) labels are designed for applications that use barcodes, alphanumerics, graphic symbols and logos, and require nameplate-like quality.
  • Matte, light grey finish gives labels a rating and serial plate appearance
  • Labels are designed to withstand numerous solvents and variable temperatures when applied to various surfaces
  • Service temperature of −40 to +120 °C (−40 to +248 °F)
Ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, and inventory and inspection labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310 for the M610/BMP61/BradyPrinter M611/M611 Label Printers or M7-R4300 for the M710/BMP71 Label Printers. This label is no longer compatible with TLS series printers.
Certificaten: UL recognised. CSA accepted. AGA Approved; Halogen-free (DIN VDE 0472/part 815).