Self-Laminating Cryogenic Polyester Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174342 311983 174347 173902 311985 174350 174371 174343 173702 173487 173905 173901 174351
BRDY174342EA 130.48 EUR
BRDY174342 BRDY311983 BRDY174347 BRDY173902 BRDY311985 BRDY174350 BRDY174371 BRDY174343 BRDY173702 BRDY173487 BRDY173905 BRDY173901 BRDY174351
Self-Laminating Cryogenic Polyester Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Etiketten
Self-laminating polyester (B-461) is a clear film with a matte white printable zone ideal for cables, laboratory ID, or general labelling where graphics need to be protected.
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance, and high/low temperature performance
  • Service temperature of −196 to +110 °C (−320 to +230 °F)
  • Matte finish
Ideal for laboratory labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printerMore Product Information